3-Amino-6-brom-2-methylpyridin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Amino-2-bromo-6-methylpyridine, 6-Bromo-2-methylpyridin-3-amine
OR40190-1G
OR40190-5G
OR40190-10G
APOSOR40190-1GEA
21.9
EUR
APOSOR40190-1G
APOSOR40190-5G
APOSOR40190-10G
3-Amino-6-brom-2-methylpyridin
3-Amino-6-bromo-2-methylpyridine
Formel:
C₆H₇BrN₂ Molecular Weight: 187.04 g/mol Schmelzpunkt: 110…111 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095086 CAS-Nummer: 126325-47-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...