3-Amino-2,6-dibrompyridin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,6-Dibromopyridin-3-amine
OR40191-25G
OR40191-5G
APOSOR40191-25GEA
62.5
EUR
APOSOR40191-25G
APOSOR40191-5G
3-Amino-2,6-dibrompyridin
3-Amino-2,6-dibromopyridine
Formel:
C₅H₄Br₂N₂ Molecular Weight: 251,9100 g/mol Schmelzpunkt: 147…148 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00128862 CAS-Nummer: 39856-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...