Piperidin-3-one hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Azacyclohexan-1-one hydrochloride, 3-Oxopiperidine hydrochloride
OR40196-5G
OR40196-1G
OR40196-250MG
APOSOR40196-5GEA
112
EUR
APOSOR40196-5G
APOSOR40196-1G
APOSOR40196-250MG
Piperidin-3-one hydrochloride
Piperidin-3-one hydrochloride
Formel:
C₅H₁₀ClNO Molecular Weight: 135,59 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD06796739 CAS-Nummer: 61644-00-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...