3-Brom-2-hydroxypyridin 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-Brom-2-pyridinol

OR40206-1G OR40206-25G OR40206-5G
APOSOR40206-1GEA 49.3 EUR
APOSOR40206-1G APOSOR40206-25G APOSOR40206-5G
3-Brom-2-hydroxypyridin 98%
3-Brom-2-hydroxypyridin
Formel: C₅H₄BrNO
Molecular Weight: 174 g/mol
Schmelzpunkt: 179…189 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03411569
CAS-Nummer: 13466-43-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR