5-Methylthiazole-4-carboxylic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR40207-1G
OR40207-250MG
OR40207-5G
APOSOR40207-1GEA
43.5
EUR
APOSOR40207-1G
APOSOR40207-250MG
APOSOR40207-5G
5-Methylthiazole-4-carboxylic acid 98%
5-Methylthiazole-4-carboxylic acid
Formel:
C5H5NO2S Molecular Weight: 143,1600 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD06738813 CAS-Nummer: 120237-76-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...