(S)-1-(tert-Butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2S)-4-Oxopiperidine-2-carboxylic acid, N-BOC protected, 4-Oxo-L-pipecolinic acid, N-BOC protected, (S)-(-)-1-Boc-4-oxopiperidine-2-carboxylic acid, (S)-1-Boc-4-oxopipecolic acid, (2S)-1-(tert-Butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid

OR40209-1G OR40209-5G
APOSOR40209-1GEA 85.7 EUR
APOSOR40209-1G APOSOR40209-5G
(S)-1-(tert-Butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid 97%
(S)-1-(tert-Butoxycarbonyl)-4-oxopiperidine-2-carboxylic acid
Formel: C₁₁H₁₇NO₅
Molecular Weight: 243,26 g/mol
Schmelzpunkt: 121…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094867
CAS-Nummer: 198646-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR