6-Brom-3-chlorisochinolin 97%

Lieferant: Apollo Scientific
Achtung

OR40220-1G OR40220-250MG
APOSOR40220-1GEA 0 EUR
APOSOR40220-1G APOSOR40220-250MG
6-Brom-3-chlorisochinolin 97%
6-Bromo-3-chloroisoquinoline
Formel: C₉H₅BrClN
Molecular Weight: 242,4997 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08703290
CAS-Nummer: 552331-06-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR