7-Brom-3-chlorisochinolin

Lieferant: Apollo Scientific
Achtung

OR40228-1G OR40228-250MG
APOSOR40228-1GEA 159 EUR
APOSOR40228-1G APOSOR40228-250MG
7-Brom-3-chlorisochinolin
7-Bromo-3-chloroisoquinoline
Formel: C₉H₅BrClN
Molecular Weight: 242,4997 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11044699
CAS-Nummer: 1029720-65-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR