1H-Indole-7-carbonitrile 97%
Lieferant: Apollo Scientific
OR40232-25G
APOSOR40232-25GEA
174
EUR
APOSOR40232-25G
1H-Indole-7-carbonitrile 97%
1H-Indole-7-carbonitrile
Formel:
C₉H₆N₂ Molecular Weight: 142,1573 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00800653 CAS-Nummer: 96631-87-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...