3-Iod-4-methylbenzonitril

Lieferant: Apollo Scientific
Achtung

OR40274-1G OR40274-5G
APOSOR40274-1GEA 21.9 EUR
APOSOR40274-1G APOSOR40274-5G
3-Iod-4-methylbenzonitril
3-Iod-4-methylbenzonitril
Formel: C₈H₆IN
Molecular Weight: 243,444 g/mol
Schmelzpunkt: 55…58 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06797817
CAS-Nummer: 42872-79-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR