Methyl-5-bromo-1,3-benzoxazole-7-carboxylate 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 5-bromobenzo[d]oxazole-7-carboxylate
OR40282-1G
OR40282-500MG
APOSOR40282-1GEA
237
EUR
APOSOR40282-1G
APOSOR40282-500MG
Methyl-5-bromo-1,3-benzoxazole-7-carboxylate 95+%
Methyl-5-bromo-1,3-benzoxazole-7-carboxylate
Formel:
C₉H₆BrNO₃ Molecular Weight: 256.06 g/mol Schmelzpunkt: 165…167 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD16140293 CAS-Nummer: 1221792-83-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...