[(1R,5S,6R)-3-Benzyl-3-azabicyclo[3.1.0]hexan-6-yl]methanol 95+%

Lieferant: Apollo Scientific

Synonyme: (1R,5S,6R)-3-Aza-3-benzyl-6-(hydroxymethyl)bicyclo[3.1.0]hexane, rel-((1R,5S,6r)-3-Benzyl-3-azabicyclo[3.1.0]hexan-6-yl)methanol

OR40298-1G
APOSOR40298-1GEA 33.5 EUR
APOSOR40298-1G
[(1R,5S,6R)-3-Benzyl-3-azabicyclo[3.1.0]hexan-6-yl]methanol 95+%
[(1R,5S,6R)-3-Benzyl-3-azabicyclo[3.1.0]hexan-6-yl]methanol
Formel: C₁₃H₁₇NO
Molecular Weight: 203.29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16140332
CAS-Nummer: 134575-07-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR