N-tert-Butyl-4-boronobenzolsulfonamid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(tert-Butylsulphamoyl)benzeneboronic acid, (4-(N-(tert-Butyl)sulphamoyl)phenyl)boronic acid
OR4030-1G
OR4030-250MG
OR4030-5G
APOSOR4030-1GEA
120
EUR
APOSOR4030-1G
APOSOR4030-250MG
APOSOR4030-5G
N-tert-Butyl-4-boronobenzolsulfonamid 97%
N-tert-Butyl-4-boronobenzolsulfonamid
Formel:
C₁₀H₁₆BNO₄S Molecular Weight: 257.11 g/mol Schmelzpunkt: 128…132 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659841 CAS-Nummer: 208516-15-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...