Methyl-4-methylisoxazole-5-carboxylate 95%

Lieferant: Apollo Scientific

OR40303-100MG OR40303-1G OR40303-250MG
APOSOR40303-100MGEA 223 EUR
APOSOR40303-100MG APOSOR40303-1G APOSOR40303-250MG
Methyl-4-methylisoxazole-5-carboxylate 95%
Methyl-4-methylisoxazole-5-carboxylate
Formel: C₆H₇NO₃
Molecular Weight: 141,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12026361
CAS-Nummer: 261350-48-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR