trans-1-Benzyl-4-(pyridin-2-yl)pyrrolidine-3-carboxylic acid 95+%
Lieferant: Apollo Scientific
OR40306-1G
OR40306-500MG
APOSOR40306-1GEA
0
EUR
APOSOR40306-1G
APOSOR40306-500MG
trans-1-Benzyl-4-(pyridin-2-yl)pyrrolidine-3-carboxylic acid 95+%
trans-1-Benzyl-4-(pyridin-2-yl)pyrrolidine-3-carboxylic acid
Formel:
C₁₇H₁₈N₂O₂ Schmelzpunkt: 85…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD16140334 CAS-Nummer: 1221791-62-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...