Methyl-4-chloro-3-methoxy-5-nitrobenzenecarboxylate 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 4-chloro-3-methoxy-5-nitrobenzoate
OR40313-1G
OR40313-5G
OR40313-25G
APOSOR40313-1GEA
21.9
EUR
APOSOR40313-1G
APOSOR40313-5G
APOSOR40313-25G
Methyl-4-chloro-3-methoxy-5-nitrobenzenecarboxylate 95+%
Methyl-4-chloro-3-methoxy-5-nitrobenzenecarboxylate
Formel:
C₉H₈ClNO₅ Molecular Weight: 245.62 g/mol Schmelzpunkt: 103…105 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD16140357 CAS-Nummer: 63603-09-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...