Methyl-2-cyano-8-methoxy-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylate 95+%
Lieferant: Apollo Scientific
OR40314-1G
OR40314-500MG
APOSOR40314-1GEA
671
EUR
APOSOR40314-1G
APOSOR40314-500MG
Methyl-2-cyano-8-methoxy-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylate 95+%
Methyl-2-cyano-8-methoxy-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylate
Formel:
C₁₂H₁₂N₂O₄ Molecular Weight: 248,24 g/mol Schmelzpunkt: 163…165 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD16140358 CAS-Nummer: 1221792-69-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...