4-Phenylcarbamoylbenzeneboronic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Phenylaminocarbonyl)benzeneboronic acid, 4-Phenylaminocarbonylphenylboronic acid, (4-(Phenylcarbamoyl)phenyl)boronic acid

OR4040-5G
APOSOR4040-5GEA 156 EUR
APOSOR4040-5G
4-Phenylcarbamoylbenzeneboronic acid 97%
4-Phenylcarbamoylbenzeneboronic acid
Formel: C₁₃H₁₂BNO₃
Molecular Weight: 241,05 g/mol
Schmelzpunkt: 210…216 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04115683
CAS-Nummer: 330793-45-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR