3-Bromo-5-methoxyaniline 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR40401-1G
OR40401-5G
OR40401-25G
APOSOR40401-1GEA
29
EUR
APOSOR40401-1G
APOSOR40401-5G
APOSOR40401-25G
3-Bromo-5-methoxyaniline 97%
3-Bromo-5-methoxyaniline
Formel:
C₇H₈BrNO Schmelzpunkt: 51…54 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11878497 CAS-Nummer: 16618-68-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...