3-Hydroxy-2-pyrrolidinone

Lieferant: Apollo Scientific
Achtung

OR40442-1G OR40442-250MG
APOSOR40442-1GEA 148 EUR
APOSOR40442-1G APOSOR40442-250MG
3-Hydroxy-2-pyrrolidinone
3-Hydroxy-2-pyrrolidinone
Formel: C₄H₇NO₂
Molecular Weight: 101.11 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09751256
CAS-Nummer: 15166-68-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR