Methyl-2-aminoisonicotinat 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Methyl 2-aminopyridine-4-carboxylate, Methyl 2-aminoisonicotinate

OR40455-25G OR40455-1G OR40455-5G
APOSOR40455-25GEA 60.9 EUR
APOSOR40455-25G APOSOR40455-1G APOSOR40455-5G
Methyl-2-aminoisonicotinat 98%
Methyl-2-aminoisonicotinat
Formel: C₇H₈N₂O₂
Molecular Weight: 152.15 g/mol
Schmelzpunkt: 144…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04039316
CAS-Nummer: 6937-03-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR