2-Aminopyridine-4-methanol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-4-pyridinylmethanol, (2-Aminopyridin-4-yl)-methanol, (2-Aminopyridin-4-yl)methanol, (2-Aminopyrid-4-yl)methanol

OR40456-25G
APOSOR40456-25GEA 116 EUR
APOSOR40456-25G
2-Aminopyridine-4-methanol 98%
2-Aminopyridine-4-methanol
Formel: C₆H₈N₂O
Molecular Weight: 124,143 g/mol
Schmelzpunkt: 80…85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03791261
CAS-Nummer: 105250-17-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR