3-Oxocyclobutancarbonsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR40464-25G
OR40464-5G
OR40464-100G
APOSOR40464-25GEA
58
EUR
APOSOR40464-25G
APOSOR40464-5G
APOSOR40464-100G
3-Oxocyclobutancarbonsäure 97%
3-Oxocyclobutanecarboxylic acid
Formel:
C5H6O3 Molecular Weight: 114,10 g/mol Schmelzpunkt: 69…70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00100900 CAS-Nummer: 23761-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...