5-Bromo-2-(piperazin-1yl)pyrimidine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-2-(1-piperazinyl)pyrimidine
OR4053-25G
OR4053-5G
APOSOR4053-25GEA
417
EUR
APOSOR4053-25G
APOSOR4053-5G
5-Bromo-2-(piperazin-1yl)pyrimidine
5-Bromo-2-(piperazin-1yl)pyrimidine
Formel:
C₈H₁₁BrN₄ Molecular Weight: 243,11 g/mol Schmelzpunkt: 77…79 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01314280 CAS-Nummer: 99931-82-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...