2,5-Dichlorbenzoxazol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,5-Dichlorobenzooxazole

OR40570-100G OR40570-25G
APOSOR40570-100GEA 156 EUR
APOSOR40570-100G APOSOR40570-25G
2,5-Dichlorbenzoxazol
2,5-Dichlorobenzoxazole
Formel: C₇H₃Cl₂NO
Molecular Weight: 188,0100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09264312
CAS-Nummer: 3621-81-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR