2-Chloro-5-hydroxypyrimidine 98%

Lieferant: Apollo Scientific
Achtung

OR40574-100G OR40574-25G OR40574-5G
APOSOR40574-100GEA 719 EUR
APOSOR40574-100G APOSOR40574-25G APOSOR40574-5G
2-Chloro-5-hydroxypyrimidine 98%
2-Chloro-5-hydroxypyrimidine
Formel: C₄H₃ClN₂O
Molecular Weight: 130,5300 g/mol
Schmelzpunkt: 195…196 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09743796
CAS-Nummer: 4983-28-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR