Dicyclohexyl(2',6'-dimethoxy-2-biphenylyl)phosphin 98%
Lieferant: Apollo Scientific
Achtung
Synonyme: 2-Dicyclohexylphosphino-2’,6’-dimethoxybiphenyl, SPhos, Dicyclohexyl(2',6'-dimethoxy-[1,1'-biphenyl]-2-yl)phosphine, 2-Dicyclohexylphosphino-2',6'-dimethoxybiphenyl, 2-Dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl
OR40584-25G OR40584-5G OR40584-100G
APOSOR40584-25GEA 34.8 EUR
APOSOR40584-25G APOSOR40584-5G APOSOR40584-100G
Dicyclohexyl(2',6'-dimethoxy-2-biphenylyl)phosphin 98%
Dicyclohexyl(2',6'-dimethoxy[1,1'-biphenyl]-2-yl)phosphine
Usage subject to US Patents 6307087 and 6395916. This product is intended for research and development purposes only.
Formel: C₂₆H₃₅O₂P
Molecular Weight: 410,53 g/mol
Schmelzpunkt: 164…167 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05861611
CAS-Nummer: 657408-07-6