4-[2-(Boc-amino)ethyl]aniline

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 4-aminophenethylcarbamate, 4-(2-Aminoethyl)aniline, 4-BOC protected

OR40588-1G
APOSOR40588-1GEA 21.9 EUR
APOSOR40588-1G
4-[2-(Boc-amino)ethyl]aniline
4-[2-(Boc-amino)ethyl]aniline
Formel: C₁₃H₂₀N₂O₂
Molecular Weight: 236.315 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD05663961
CAS-Nummer: 94838-59-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR