2-Amino-4-chlor-3-nitropyridin

Lieferant: Apollo Scientific
Achtung

APOSOR40607-1GEA 14.7 EUR
APOSOR40607-1G APOSOR40607-5G
2-Amino-4-chlor-3-nitropyridin
2-Amino-4-chloro-3-nitropyridine
Formel: C₅H₄ClN₃O₂
Molecular Weight: 173,56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04116097
CAS-Nummer: 6980-08-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR