3-Nitrophthalonitril 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Nitrophthalsäuredinitril, 3-Nitrobenzol-1,2-dicarbonitril

OR40623-100G OR40623-25G OR40623-500G
APOSOR40623-100GEA 45.1 EUR
APOSOR40623-100G APOSOR40623-25G APOSOR40623-500G
3-Nitrophthalonitril 99%
3-Nitrophthalonitril
Formel: C₈H₃N₃O₂
Molecular Weight: 173,13 g/mol
Schmelzpunkt: 162…165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191558
CAS-Nummer: 51762-67-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR