5-Brom-2-iodbenzaldehyd

Lieferant: Apollo Scientific
Achtung

OR40680-1G OR40680-5G OR40680-10G
APOSOR40680-1GEA 21.9 EUR
APOSOR40680-1G APOSOR40680-5G APOSOR40680-10G
5-Brom-2-iodbenzaldehyd
5-Brom-2-iodbenzaldehyd
Formel: C₇H₄BrIO
Molecular Weight: 310,9200 g/mol
Schmelzpunkt: 89…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07779026
CAS-Nummer: 689291-89-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR