4-Brom-3-iodanisol 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-2-iodo-4-methoxybenzene
OR40681-1G
OR40681-250MG
OR40681-5G
APOSOR40681-1GEA
50.9
EUR
APOSOR40681-1G
APOSOR40681-250MG
APOSOR40681-5G
4-Brom-3-iodanisol 95%
4-Brom-3-iodanisol
Formel:
C₇H₆BrIO Molecular Weight: 312,9300 g/mol Siedepunkt: 124…126 °C/2mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00070743 CAS-Nummer: 4897-68-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...