6-Chlorisatin 97+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Coroindoline-2,3-dione

OR40689-100G OR40689-25G
APOSOR40689-100GEA 249 EUR
APOSOR40689-100G APOSOR40689-25G
6-Chlorisatin 97+%
6-Chlorisatin
Formel: C₈H₄ClNO₂
Molecular Weight: 181,58 g/mol
Schmelzpunkt: 262 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00086347
CAS-Nummer: 6341-92-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR