2-Chlor-3-nitro-5-thiophensulfonylchlorid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-Chloro-4-nitrothiophene-2-sulphonyl chloride

OR4069-1G
APOSOR4069-1GEA 29 EUR
APOSOR4069-1G
2-Chlor-3-nitro-5-thiophensulfonylchlorid
2-Chlor-3-nitro-5-thiophensulfonylchlorid
Formel: C₄HCl₂NO₄S₂
Molecular Weight: 262.09 g/mol
Schmelzpunkt: 51…54 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00067978
CAS-Nummer: 58457-24-2
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR