2-Brom-5-methyl-1,3,4-thiadiazol 95%

Lieferant: Apollo Scientific
Achtung

OR40731-5G OR40731-1G
APOSOR40731-5GEA 69.6 EUR
APOSOR40731-5G APOSOR40731-1G
2-Brom-5-methyl-1,3,4-thiadiazol 95%
2-Bromo-5-methyl-1,3,4-thiadiazole
Formel: C₃H₃BrN₂S
Molecular Weight: 179,04 g/mol
Schmelzpunkt: 105…113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00518965
CAS-Nummer: 54044-79-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR