6-Chloro-1H-indazole-3-carboxylic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR40751-10G
OR40751-1G
OR40751-5G
APOSOR40751-10GEA
519
EUR
APOSOR40751-10G
APOSOR40751-1G
APOSOR40751-5G
6-Chloro-1H-indazole-3-carboxylic acid 97%
6-Chloro-1H-indazole-3-carboxylic acid
Formel:
C₈H₅ClN₂O₂ Molecular Weight: 196,5900 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05663998 CAS-Nummer: 129295-31-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...