1,2,3,4-Tetrahydroisoquinoline-1-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,2,3,4-Tetrahydroisoquinoline-1-carboxylic acid
OR40776-1G
OR40776-5G
APOSOR40776-1GEA
58
EUR
APOSOR40776-1G
APOSOR40776-5G
1,2,3,4-Tetrahydroisoquinoline-1-carboxylic acid
1,2,3,4-Tetrahydroisoquinoline-1-carboxylic acid
Formel:
C₁₀H₁₁NO₂ Molecular Weight: 177,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01318704 CAS-Nummer: 41034-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...