3-(4-Chlorphenoxy)propionsäure 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(4-Chlorophenoxy)propanoic acid
OR40783-25G
OR40783-5G
APOSOR40783-25GEA
105
EUR
APOSOR40783-25G
APOSOR40783-5G
3-(4-Chlorphenoxy)propionsäure 99%
3-(4-Chlorphenoxy)propionsäure
Formel:
C₉H₉ClO₃ Molecular Weight: 200,6190 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00043504 CAS-Nummer: 3284-79-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...