Dirhodium(II) tetrakis (caprolactam) 98%

Lieferant: Apollo Scientific

Synonyme: Dirhodium tetracaprolactamate

OR40791-100MG OR40791-250MG OR40791-1G
APOSOR40791-100MGEA 229 EUR
APOSOR40791-100MG APOSOR40791-250MG APOSOR40791-1G
Dirhodium(II) tetrakis (caprolactam) 98%
Dirhodium(II) tetrakis (caprolactam)
Formel: C₂₄H₄₀N₄O₄Rh₂
Molecular Weight: 654,419 g/mol
Schmelzpunkt: 269…274 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00467690
CAS-Nummer: 138984-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR