2-(Methylthio)thiophen
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Mehtylthio)thiophene
OR4091-10G
OR4091-5G
OR4091-1G
APOSOR4091-10GEA
163
EUR
APOSOR4091-10G
APOSOR4091-5G
APOSOR4091-1G
2-(Methylthio)thiophen
2-(Methylthio)thiophene
Formel:
C₅H₆S₂ Molecular Weight: 130.23 g/mol Siedepunkt: 82…84 °C/15mmHg Lagertemperatur: Kühlschrank |
MDL:
MFCD00052382 CAS-Nummer: 5780-36-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...