2-Acetamido-5-aminopyridin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Acetylamino)-5-aminopyridine

OR4098-25G OR4098-5G
APOSOR4098-25GEA 427 EUR
APOSOR4098-25G APOSOR4098-5G
2-Acetamido-5-aminopyridin 98%
2-Acetamido-5-aminopyridin
Formel: C₇H₉N₃O
Molecular Weight: 151,17 g/mol
Schmelzpunkt: 154 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00160311
CAS-Nummer: 29958-14-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR