6-Chlor-5-nitrochinolin 98%
Lieferant: Apollo Scientific
OR41004-5G
APOSOR41004-5GEA
79.9
EUR
APOSOR41004-5G
6-Chlor-5-nitrochinolin 98%
6-Chloro-5-nitroquinoline
Formel:
C₉H₅ClN₂O₂ Molecular Weight: 208,6012 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00093616 CAS-Nummer: 86984-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...