4-Chlor-6-methylchinolin

Lieferant: Apollo Scientific

OR41005-5G
APOSOR41005-5GEA 79.9 EUR
APOSOR41005-5G
4-Chlor-6-methylchinolin
4-Chloro-6-methylquinoline
Formel: C₁₀H₈ClN
Molecular Weight: 177,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02684204
CAS-Nummer: 18436-71-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR