3-Methylmorpholine 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR41010-5G
APOSOR41010-5GEA
58
EUR
APOSOR41010-5G
3-Methylmorpholine 97%
3-Methylmorpholine
Formel:
C5H11NO Molecular Weight: 101,1469 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09054773 CAS-Nummer: 42185-06-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...