3-Brom-9-phenyl-9H-carbazol 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-Bromo-9-phenylcarbazole

OR41026-100G
APOSOR41026-100GEA 102 EUR
APOSOR41026-100G
3-Brom-9-phenyl-9H-carbazol 98%
3-Bromo-9-phenyl-9H-carbazole
Formel: C18H12BrN
Molecular Weight: 322,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11977305
CAS-Nummer: 1153-85-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR