1-Adamantyl methacrylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Methacryloyloxyadamantane, Methacrylic acid 1-Adamantyl ester

OR41031-100G OR41031-25G
APOSOR41031-100GEA 482 EUR
APOSOR41031-100G APOSOR41031-25G
1-Adamantyl methacrylate
1-Adamantyl methacrylate
Formel: C₁₄H₂₀O₂
Molecular Weight: 220,31 g/mol
Siedepunkt: 113 °C/3mmHg
Dichte: 1,0495 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD08703330
CAS-Nummer: 16887-36-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR