5-Brom-o-anisidin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-4-bromanisol, 5-Brom-2-anisidin, 5-Brom-2-methoxyanilin
OR41052-25G
OR41052-5G
APOSOR41052-25GEA
120
EUR
APOSOR41052-25G
APOSOR41052-5G
5-Brom-o-anisidin 98%
5-Brom-o-anisidin
Formel:
C₇H₈BrNO Molecular Weight: 202,05 g/mol Schmelzpunkt: 94…96 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04037882 CAS-Nummer: 6358-77-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...