Methyl-4-azaindole-2-carboxylate
Lieferant: Apollo Scientific
OR41118-1G
OR41118-250MG
APOSOR41118-1GEA
473
EUR
APOSOR41118-1G
APOSOR41118-250MG
Methyl-4-azaindole-2-carboxylate
Methyl-4-azaindole-2-carboxylate
Formel:
C₉H₈N₂O₂ Molecular Weight: 176,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09743416 CAS-Nummer: 394223-19-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...