5-(Aminomethyl)pyridin-2-amine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-5-(aminomethyl)pyridine
OR41126-1G
OR41126-250MG
OR41126-500MG
APOSOR41126-1GEA
481
EUR
APOSOR41126-1G
APOSOR41126-250MG
APOSOR41126-500MG
5-(Aminomethyl)pyridin-2-amine
5-(Aminomethyl)pyridin-2-amine
Formel:
C₆H₉N₃ Molecular Weight: 123,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06213156 CAS-Nummer: 156973-09-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...