6-Chloropyrimidine-4-carbonitrile 97%

Lieferant: Apollo Scientific

OR41172-1G OR41172-5G
APOSOR41172-1GEA 186 EUR
APOSOR41172-1G APOSOR41172-5G
6-Chloropyrimidine-4-carbonitrile 97%
6-Chloropyrimidine-4-carbonitrile
Formel: C₅H₂ClN₃
Molecular Weight: 139,54 g/mol
Schmelzpunkt: 25 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09607731
CAS-Nummer: 939986-65-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR